Anti-NODAL Rat Monoclonal Antibody [clone: 209009]

Supplier: R & D Systems
MAB1315
220018-986EA 772.25 USD
220018-986
Anti-NODAL Rat Monoclonal Antibody [clone: 209009]
Antibodies
Type: Primary
Antigen: NODAL
Clonality: Monoclonal
Clone: 209009
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR