Anti-LGALS8 Mouse Monoclonal Antibody [clone: 210608]

Supplier: R & D Systems
MAB1305
220018-982EA 772.25 USD
220018-982
Anti-LGALS8 Mouse Monoclonal Antibody [clone: 210608]
Antibodies
Type: Primary
Antigen: LGALS8
Clonality: Monoclonal
Clone: 210608
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR