Anti-LGALS7 Rat Monoclonal Antibody [clone: 212923]

Supplier: R & D Systems
MAB1304
220018-981EA 763.57 USD
220018-981
Anti-LGALS7 Rat Monoclonal Antibody [clone: 212923]
Antibodies
Type: Primary
Antigen: LGALS7
Clonality: Monoclonal
Clone: 212923
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR