Anti-LY86 Rat Monoclonal Antibody [clone: 142004]

Supplier: R & D Systems
MAB130
220018-977EA 763.57 USD
220018-977
Anti-LY86 Rat Monoclonal Antibody [clone: 142004]
Antibodies
Type: Primary
Antigen: LY86
Clonality: Monoclonal
Clone: 142004
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR