Anti-CST4 Mouse Monoclonal Antibody [clone: 201805]

Supplier: R & D Systems
MAB1296
220018-975EA 772.25 USD
220018-975
Anti-CST4 Mouse Monoclonal Antibody [clone: 201805]
Antibodies
Type: Primary
Antigen: CST4
Clonality: Monoclonal
Clone: 201805
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR