Anti-SERPINA3 Mouse Monoclonal Antibody [clone: 213907]

Supplier: R & D Systems
MAB1295
220018-974EA 763.57 USD
220018-974
Anti-SERPINA3 Mouse Monoclonal Antibody [clone: 213907]
Antibodies
Type: Primary
Antigen: SERPINA3
Clonality: Monoclonal
Clone: 213907
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR