Anti-CTSE Mouse Monoclonal Antibody [clone: 212211]

Supplier: R & D Systems
MAB1294
220018-973EA 763.57 USD
220018-973
Anti-CTSE Mouse Monoclonal Antibody [clone: 212211]
Antibodies
Type: Primary
Antigen: CTSE
Clonality: Monoclonal
Clone: 212211
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR