Anti-CST6 Mouse Monoclonal Antibody [clone: 211515]

Supplier: R & D Systems
MAB1286
220018-970EA 772.25 USD
220018-970
Anti-CST6 Mouse Monoclonal Antibody [clone: 211515]
Antibodies
Type: Primary
Antigen: CST6
Clonality: Monoclonal
Clone: 211515
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR