Anti-CST1 Mouse Monoclonal Antibody [clone: 213139]

Supplier: R & D Systems
MAB1285
220018-969EA 772.25 USD
220018-969
Anti-CST1 Mouse Monoclonal Antibody [clone: 213139]
Antibodies
Type: Primary
Antigen: CST1
Clonality: Monoclonal
Clone: 213139
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR