Anti-CST6 Rat Monoclonal Antibody [clone: 216820]

Supplier: R & D Systems
MAB1284
220018-968EA 763.57 USD
220018-968
Anti-CST6 Rat Monoclonal Antibody [clone: 216820]
Antibodies
Type: Primary
Antigen: CST6
Clonality: Monoclonal
Clone: 216820
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR