Anti-CD72 Rat Monoclonal Antibody [clone: 164726]

Supplier: R & D Systems
MAB1279
220018-966EA 763.57 USD
220018-966
Anti-CD72 Rat Monoclonal Antibody [clone: 164726]
Antibodies
Type: Primary
Antigen: CD72
Clonality: Monoclonal
Clone: 164726
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR