Anti-SLPI Mouse Monoclonal Antibody [clone: 20409]

Supplier: R & D Systems
MAB1274
220018-960EA 763.57 USD
220018-960
Anti-SLPI Mouse Monoclonal Antibody [clone: 20409]
Antibodies
Type: Primary
Antigen: SLPI
Clonality: Monoclonal
Clone: 20409
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR