Anti-DKK4 Mouse Monoclonal Antibody [clone: 185709]

Supplier: R & D Systems
MAB1269-050 MAB1269-500
220018-957EA 339.85 USD
220018-957 220018-958
Anti-DKK4 Mouse Monoclonal Antibody [clone: 185709]
Antibodies
Type: Primary
Antigen: DKK4
Clonality: Monoclonal
Clone: 185709
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR