Anti-SERPINA5 Mouse Monoclonal Antibody [clone: 210326]

Supplier: R & D Systems
MAB1266
220018-955EA 763.57 USD
220018-955
Anti-SERPINA5 Mouse Monoclonal Antibody [clone: 210326]
Antibodies
Type: Primary
Antigen: SERPINA5
Clonality: Monoclonal
Clone: 210326
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR