Anti-PDGFRB Mouse Monoclonal Antibody [clone: PR7212]

Supplier: R & D Systems
MAB1263
220018-953EA 588.58 USD
220018-953
Anti-PDGFRB Mouse Monoclonal Antibody [clone: PR7212]
Antibodies
Type: Primary
Antigen: PDGFRB
Clonality: Monoclonal
Clone: PR7212
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR