Anti-SLC2A4 Mouse Monoclonal Antibody [clone: 1F8]

Supplier: R & D Systems
MAB1262
220018-952EA 616.06 USD
220018-952
Anti-SLC2A4 Mouse Monoclonal Antibody [clone: 1F8]
Antibodies
Immunoaffinity Purification

Type: Primary
Antigen: SLC2A4
Clonality: Monoclonal
Clone: 1F8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR