Anti-FASLG Mouse Monoclonal Antibody [clone: 154922]

Supplier: R & D Systems
MAB1261
220018-951EA 763.57 USD
220018-951
Anti-FASLG Mouse Monoclonal Antibody [clone: 154922]
Antibodies
Type: Primary
Antigen Symbol: FASLG
Antigen Name: Fas ligand (TNF superfamily, member 6)
Antigen Synonyms: ALPS1B, APT1LG1, APTL, CD178, CD95-L, CD95L, FASL, TNFSF6
Clonality: Monoclonal
Clone: 154922
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR