Anti-NES Mouse Monoclonal Antibody [clone: 196908]

Supplier: R & D Systems
MAB1259
220018-948EA 655.11 USD
220018-948
Anti-NES Mouse Monoclonal Antibody [clone: 196908]
Antibodies
Type: Primary
Antigen Symbol: NES
Antigen Name: nestin
Antigen Synonyms: Nbla00170
Clonality: Monoclonal
Clone: 196908
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR