Anti-FCGR1A Mouse Monoclonal Antibody [clone: 276426]

Supplier: R & D Systems
MAB12571
220018-946EA 594.37 USD
220018-946
Anti-FCGR1A Mouse Monoclonal Antibody [clone: 276426]
Antibodies
Type: Primary
Antigen Symbol: FCGR1A
Antigen Name: Fc fragment of IgG, high affinity Ia, receptor (CD64)
Antigen Synonyms: CD64, CD64A, FCRI, IGFR1
Clonality: Monoclonal
Clone: 276426
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR