Anti-FSTL3 Rat Monoclonal Antibody [clone: 210725]

Supplier: R & D Systems
MAB1255
220018-944EA 763.57 USD
220018-944
Anti-FSTL3 Rat Monoclonal Antibody [clone: 210725]
Antibodies
Type: Primary
Antigen Symbol: FSTL3
Antigen Name: follistatin-like 3
Antigen Synonyms: E030038F23Rik, Flrg
Clonality: Monoclonal
Clone: 210725
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR