Anti-NTN4 Mouse Monoclonal Antibody [clone: 191633]

Supplier: R & D Systems
MAB1254
220018-943EA 763.57 USD
220018-943
Anti-NTN4 Mouse Monoclonal Antibody [clone: 191633]
Antibodies
Type: Primary
Antigen: NTN4
Clonality: Monoclonal
Clone: 191633
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR