Anti-SEMA3A Mouse Monoclonal Antibody [clone: 215803]

Supplier: R & D Systems
MAB1250
220018-942EA 647.88 USD
220018-942
Anti-SEMA3A Mouse Monoclonal Antibody [clone: 215803]
Antibodies
Type: Primary
Antigen: SEMA3A
Clonality: Monoclonal
Clone: 215803
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR