Anti-GCG Mouse Monoclonal Antibody [clone: 181402]

Supplier: R & D Systems
MAB1249
220018-941EA 594.37 USD
220018-941
Anti-GCG Mouse Monoclonal Antibody [clone: 181402]
Antibodies
Type: Primary
Antigen Symbol: GCG
Antigen Name: glucagon
Antigen Synonyms: GLP-1, Glu, PPG
Clonality: Monoclonal
Clone: 181402
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR