Anti-VEGF Mouse Monoclonal Antibody [clone: 211615]

Supplier: R & D Systems
MAB1247
220018-939EA 763.57 USD
220018-939
Anti-VEGF Mouse Monoclonal Antibody [clone: 211615]
Antibodies
Type: Primary
Antigen: VEGF
Clonality: Monoclonal
Clone: 211615
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Zebrafish
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR