Anti-LGALS1 Rat Monoclonal Antibody [clone: 201002]

Supplier: R & D Systems
MAB12451
220018-937EA 772.25 USD
220018-937
Anti-LGALS1 Rat Monoclonal Antibody [clone: 201002]
Antibodies
Type: Primary
Antigen: LGALS1
Clonality: Monoclonal
Clone: 201002
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR