Anti-LGALS1 Rat Monoclonal Antibody [clone: 201066]

Supplier: R & D Systems
MAB1245
220018-936EA 763.57 USD
220018-936
Anti-LGALS1 Rat Monoclonal Antibody [clone: 201066]
Antibodies
Type: Primary
Antigen: LGALS1
Clonality: Monoclonal
Clone: 201066
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR