Anti-MDM2 Mouse Monoclonal Antibody [clone: 197702]

Supplier: R & D Systems
MAB1244
220018-935EA 594.37 USD
220018-935
Anti-MDM2 Mouse Monoclonal Antibody [clone: 197702]
Antibodies
Type: Primary
Antigen: MDM2
Clonality: Monoclonal
Clone: 197702
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR