Anti-TNFSF13B Mouse Monoclonal Antibody [clone: 137314]

Supplier: R & D Systems
MAB1241
220018-934EA 772.25 USD
220018-934
Anti-TNFSF13B Mouse Monoclonal Antibody [clone: 137314]
Antibodies
Type: Primary
Antigen: TNFSF13B
Clonality: Monoclonal
Clone: 137314
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR