Anti-IGFBP1 Rat Monoclonal Antibody [clone: 176110]

Supplier: R & D Systems
MAB1240
220018-933EA 763.57 USD
220018-933
Anti-IGFBP1 Rat Monoclonal Antibody [clone: 176110]
Antibodies
Type: Primary
Antigen: IGFBP1
Clonality: Monoclonal
Clone: 176110
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR