Anti-SERPINF2 Rat Monoclonal Antibody [clone: 209126]

Supplier: R & D Systems
MAB1239
220018-931EA 772.25 USD
220018-931
Anti-SERPINF2 Rat Monoclonal Antibody [clone: 209126]
Antibodies
Type: Primary
Antigen: SERPINF2
Clonality: Monoclonal
Clone: 209126
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR