Anti-CST3 Rat Monoclonal Antibody [clone: 207103]

Supplier: R & D Systems
MAB1238
220018-930EA 763.57 USD
220018-930
Anti-CST3 Rat Monoclonal Antibody [clone: 207103]
Antibodies
Type: Primary
Antigen: CST3
Clonality: Monoclonal
Clone: 207103
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR