Anti-TNFSF12 Rat Monoclonal Antibody [clone: 189803]

Supplier: R & D Systems
MAB1237
220018-929EA 763.57 USD
220018-929
Anti-TNFSF12 Rat Monoclonal Antibody [clone: 189803]
Antibodies
Type: Primary
Antigen: TNFSF12
Clonality: Monoclonal
Clone: 189803
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR