Anti-NTM Mouse Monoclonal Antibody [clone: 175214]

Supplier: R & D Systems
MAB1235
220018-926EA 763.57 USD
220018-926
Anti-NTM Mouse Monoclonal Antibody [clone: 175214]
Antibodies
Type: Primary
Antigen: NTM
Clonality: Monoclonal
Clone: 175214
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR