Anti-IL17C Mouse Monoclonal Antibody [clone: 177114]

Supplier: R & D Systems
MAB1234
220018-925EA 779.48 USD
220018-925
Anti-IL17C Mouse Monoclonal Antibody [clone: 177114]
Antibodies
Type: Primary
Antigen: IL17C
Clonality: Monoclonal
Clone: 177114
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR