Anti-ITGA2 Rat Monoclonal Antibody [clone: 430903]

Supplier: R & D Systems
MAB12332
220018-924EA 577.02 USD
220018-924
Anti-ITGA2 Rat Monoclonal Antibody [clone: 430903]
Antibodies
Type: Primary
Antigen: ITGA2
Clonality: Monoclonal
Clone: 430903
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR