Anti-ITGA2 Mouse Monoclonal Antibody [clone: HAS3]

Supplier: R & D Systems
MAB1233
220018-922EA 779.48 USD
220018-922
Anti-ITGA2 Mouse Monoclonal Antibody [clone: HAS3]
Antibodies
Type: Primary
Antigen: ITGA2
Clonality: Monoclonal
Clone: HAS3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR