Anti-FCER2 Mouse Monoclonal Antibody [clone: 138633]

Supplier: R & D Systems
MAB1231
220018-921EA 763.57 USD
220018-921
Anti-FCER2 Mouse Monoclonal Antibody [clone: 138633]
Antibodies
Type: Primary
Antigen Symbol: FCER2
Antigen Name: Fc fragment of IgE, low affinity II, receptor for (CD23)
Antigen Synonyms: BLAST-2, CD23, CD23A, CLEC4J, FCE2, IGEBF
Clonality: Monoclonal
Clone: 138633
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR