Anti-MAPK1 Mouse Monoclonal Antibody [clone: 191801]

Supplier: R & D Systems
MAB1230
220018-920EA 600.15 USD
220018-920
Anti-MAPK1 Mouse Monoclonal Antibody [clone: 191801]
Antibodies
Simple Western

Type: Primary
Antigen: MAPK1
Clonality: Monoclonal
Clone: 191801
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR