Anti-ADAM19 Rat Monoclonal Antibody [clone: 164646]

Supplier: R & D Systems
MAB1229
220018-918EA 763.57 USD
220018-918
Anti-ADAM19 Rat Monoclonal Antibody [clone: 164646]
Antibodies
Type: Primary
Antigen: ADAM19
Clonality: Monoclonal
Clone: 164646
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR