Anti-LY6A Rat Monoclonal Antibody [clone: 177228]

Supplier: R & D Systems
MAB1226
220018-915EA 763.57 USD
220018-915
Anti-LY6A Rat Monoclonal Antibody [clone: 177228]
Antibodies
Type: Primary
Antigen: LY6A
Clonality: Monoclonal
Clone: 177228
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR