Anti-KIR3DL1 Mouse Monoclonal Antibody [clone: 177407]

Supplier: R & D Systems
MAB12251
220018-914EA 772.25 USD
220018-914
Anti-KIR3DL1 Mouse Monoclonal Antibody [clone: 177407]
Antibodies
Type: Primary
Antigen: KIR3DL1
Clonality: Monoclonal
Clone: 177407
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR