Anti-KIR3DL1 Mouse Monoclonal Antibody [clone: DX9]

Supplier: R & D Systems
MAB1225
220018-913EA 779.48 USD
220018-913
Anti-KIR3DL1 Mouse Monoclonal Antibody [clone: DX9]
Antibodies
Blockade of Receptor-ligand Interaction

Type: Primary
Antigen: KIR3DL1
Clonality: Monoclonal
Clone: DX9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR