Anti-ACAN (G1-IGD-G2 Domains) Mouse Monoclonal Antibody [clone: 17959]

Supplier: R & D Systems
MAB1220
220018-911EA 779.48 USD
220018-911
Anti-ACAN (G1-IGD-G2 Domains) Mouse Monoclonal Antibody [clone: 17959]
Antibodies
Type: Primary
Antigen: ACAN (G1-IGD-G2 Domains)
Clonality: Monoclonal
Clone: 17959
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR