Anti-PSEN2 Mouse Monoclonal Antibody [clone: 149903]

Supplier: R & D Systems
MAB1218
220018-908EA 763.57 USD
220018-908
Anti-PSEN2 Mouse Monoclonal Antibody [clone: 149903]
Antibodies
Type: Primary
Antigen: PSEN2
Clonality: Monoclonal
Clone: 149903
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR