Anti-IL18RAP Rat Monoclonal Antibody [clone: 112614]

Supplier: R & D Systems
MAB1216
220018-905EA 763.57 USD
220018-905
Anti-IL18RAP Rat Monoclonal Antibody [clone: 112614]
Antibodies
Type: Primary
Antigen: IL18RAP
Clonality: Monoclonal
Clone: 112614
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR