Anti-CD80 Mouse Monoclonal Antibody [clone: 210408]

Supplier: R & D Systems
MAB1214
220018-904EA 763.57 USD
220018-904
Anti-CD80 Mouse Monoclonal Antibody [clone: 210408]
Antibodies
Type: Primary
Antigen: CD80
Clonality: Monoclonal
Clone: 210408
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR