Anti-FGF16 Mouse Monoclonal Antibody [clone: 209511]

Supplier: R & D Systems
MAB1212
220018-903EA 763.57 USD
220018-903
Anti-FGF16 Mouse Monoclonal Antibody [clone: 209511]
Antibodies
Type: Primary
Antigen Symbol: FGF16
Antigen Name: fibroblast growth factor 16
Antigen Synonyms: FGF-16, MF4
Clonality: Monoclonal
Clone: 209511
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR