Anti-GHR Mouse Monoclonal Antibody [clone: 198314]

Supplier: R & D Systems
MAB1211
220018-902EA 763.57 USD
220018-902
Anti-GHR Mouse Monoclonal Antibody [clone: 198314]
Antibodies
Type: Primary
Antigen Symbol: GHR
Antigen Name: growth hormone receptor
Antigen Synonyms: GHR/BP
Clonality: Monoclonal
Clone: 198314
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR