Anti-GHR Mouse Monoclonal Antibody [clone: 193804]

Supplier: R & D Systems
MAB1210
220018-901EA 763.57 USD
220018-901
Anti-GHR Mouse Monoclonal Antibody [clone: 193804]
Antibodies
Type: Primary
Antigen Symbol: GHR
Antigen Name: growth hormone receptor
Antigen Synonyms: GHBP, GHIP
Clonality: Monoclonal
Clone: 193804
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR