Anti-PROK1 Mouse Monoclonal Antibody [clone: 188608]

Supplier: R & D Systems
MAB12091
220018-900EA 763.57 USD
220018-900
Anti-PROK1 Mouse Monoclonal Antibody [clone: 188608]
Antibodies
Type: Primary
Antigen: PROK1
Clonality: Monoclonal
Clone: 188608
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR